accessibility menu, dialog, popup

UserName

CAS RN 97674-02-7

CH3SnH3CCH3H2COH3C

CAS RN 97674-02-7

IUPAC Name: tributyl(1-ethoxyethenyl)stannane
Synonyms: tributyl(1-ethoxyethenyl)stannane
Molecular Weight (g/mol): 361.16
Molecular Formula: C16H34OSn
InChi Key: HGXJOXHYPGNVNK-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C(=C)OCC
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)