missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethylene Glycol (Ethylene Glycol), TRC

Product Code. 30003797
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Unit Size:
1mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003797 1 mL 1mL
30005996 25 mL 25mL
30009727 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003797 Supplier Toronto Research Chemicals Supplier No. TRCE8901401ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2 H6 O2
Molecular Weight (g/mol) 62.07
Synonym Ethylene Glycol, Ethan-1,2-diol, 1,2-Ethanediol (9CI, ACI), Ethylene glycol (8CI), Glycol (6CI, 7CI), 1,2-Dihydroxyethane, 1,2-Ethylene glycol, 146AR, 2-Hydroxyethanol, Dowtherm SR 1, E 0105, E 600, E 600 (glycol), Ethylene alcohol, Ethylene dihydrate, Fridex, Glycol alcohol, Glysantin G 34, Glysil GS, Macrogol 400 BPC, MEG 100, MeSH ID: D019855, Monoethylene glycol, Norkool, NSC 93876, Ramp, SR 01, Tescol, Ucar 17, Union Carbide XL 54 Type I De-icing Fluid, Zerex
IUPAC Name ethane-1,2-diol
SMILES OCCO

Specifications

Quantity 1 mL
InChI Formula InChI=1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
Formula Weight 62.0368
Chemical Name or Material Ethylene glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.