missing translation for 'onlineSavingsMsg'
Learn More

1,3-Divinyltetramethyldisiloxane, 96%, cont. up to 4% 1-vinyl-3-ethyltetramethyldisiloxane

Product Code. p-7020485
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11459703

Brand: Thermo Scientific Alfa Aesar A12463.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2627-95-4 | C8H18OSi2 | 186.40 g/mol

1,3-Divinyltetramethyldisiloxane acts as a potential vinyl donor used in cross-coupling reactions. It is also involved in the copolymerization reaction with aromatic ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Divinyltetramethyldisiloxane acts as a potential vinyl donor used in cross-coupling reactions. It is also involved in the copolymerization reaction with aromatic ketones.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-95-4
Molecular Formula C8H18OSi2
Molecular Weight (g/mol) 186.40
MDL Number MFCD00014933
InChI Key BITPLIXHRASDQB-UHFFFAOYSA-N
Synonym 1,3-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, 1,1'-divinyltetramethyldisiloxane, unii-sj4d71zqf4, bisvinyltetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane
PubChem CID 75819
IUPAC Name ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane
SMILES C[Si](C)(O[Si](C)(C)C=C)C=C

Specifications

Boiling Point 139°C to 140°C
Melting Point -100°C
Quantity 25 g
UN Number UN1993
Beilstein 1762585
Flash Point 24°C (75°F)
Solubility Information Immiscible with water.
Formula Weight 186.4
Percent Purity 96%
Refractive Index 1.411
Sensitivity Moisture sensitive
Density 0.811
Assay cont. up to 4% 1-vinyl-3-ethyltetramethyldisiloxane
Chemical Name or Material 1,3-Divinyltetramethyldisiloxane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.