Learn More
Fisherbrand™ 10-17mm Tube Rack
Product Code. 15728177 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
Tubes (40)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15728177 Tubes (40) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15728177
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Incorporates two rotatable, hinged shelves to accommodate varying tube size. The multi-purpose Fisherbrand™ 10-17mm tube facilitates more comfortable pipetting and makes efficient use of limited space.
  • The 10-17 mm Tube Rack comes with FOUR interchangeable tube segments.
  • Ergonomic handles for ease of transporting the rack.
  • Molded grid on both sides of the shelves for quick sample identification and retrieval.
Specifications
Autoclavable Autoclavable
Color Purple, Natural
Holds Tubes (40)
Diameter (Metric) Well 17 mm
Height (Metric) 7.7 cm
Width (Metric) 10.7 cm