missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, AISI 4330, K23080, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Alloy Steel, AISI 4330, K23080, 100g
Specifications
Specifications
| Chemical Name or Material | Niobium/Tin/Phosphorus/Cobalt/Aluminium/Tungsten/Vanadium/Copper/Silicon/Carbon/Molybdenum/Chromium/Manganese/Nickel |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Niobium 0.002 %/Tin 0.0047 %/Phosphorus 0.0075 %/Cobalt 0.013 %/Aluminium 0.023 %/Tungsten 0.024 %/Vanadium 0.076 %/Copper 0.084 %/Silicon 0.277 %/Carbon 0.302 %/Molybdenum 0.406 %/Chromium 0.807 %/Manganese 0.82 %/Nickel 1.856 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction