missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1018, G10180, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Carbon Steel, AISI 1018, G10180, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Calcium/Niobium/Titanium/Zirconium/Aluminium/Arsenic/Vanadium/Cobalt/Oxygen/Phosphorus/Tin/Nitrogen/Tungsten/Molybdenum/Nickel/Chromium/Carbon/Silicon/Copper/Manganese/Lead/Antimony |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0001 %/Calcium 0.0004 %/Niobium 0.002 %/Titanium 0.002 %/Zirconium 0.002 %/Aluminium 0.003 %/Arsenic 0.006 %/Vanadium 0.006 %/Cobalt 0.01 %/Oxygen 0.0106 %/Phosphorus 0.011 %/Tin 0.011 %/Nitrogen 0.0113 %/Tungsten 0.012 %/Molybdenum 0.031 %/Nickel 0.099 %/Chromium 0.15 %/Carbon 0.2 %/Silicon 0.22 %/Copper 0.247 %/Manganese 0.69 %/Lead (<0.005) %/Antimony (<0.01) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction