missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, 70-30 Copper Nickel, CDA 964, C96400, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, 70-30 Copper Nickel, CDA 964, C96400, 100g
Specifications
Specifications
| Chemical Name or Material | Nitrogen/Chromium/Oxygen/Zinc/Aluminium/Bismuth/Phosphorus/Cobalt/Lead/Tin/Carbon/Silicon/Iron/Niobium/Manganese/Nickel/Copper/Antimony/Titanium/Boron |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Nitrogen 0.0002 %/Chromium 0.002 %/Oxygen 0.002 %/Zinc 0.002 %/Aluminium 0.003 %/Bismuth 0.003 %/Phosphorus 0.003 %/Cobalt 0.004 %/Lead 0.004 %/Tin 0.005 %/Carbon 0.01 %/Silicon 0.19 %/Iron 0.91 %/Niobium 0.92 %/Manganese 1.04 %/Nickel 30 %/Copper 66.7 %/Antimony <0.005 %/Titanium <0.005 %/Boron <0.007 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction