missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Manganese Bronze A, CDA 675, C67500, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Manganese Bronze A, CDA 675, C67500, 100g
Specifications
Specifications
| Chemical Name or Material | Aluminium/Carbon/Phosphorus/Nickel/Lead/Manganese/Tin/Iron/Zinc/Copper/Magnesium/Oxygen/Silver/Cadmium/Chromium/Cobalt/Silicon/Antimony |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.002 %/Carbon 0.003 %/Phosphorus 0.004 %/Nickel 0.01 %/Lead 0.017 %/Manganese 0.13 %/Tin 0.85 %/Iron 0.97 %/Zinc 39.3 %/Copper 58.7 %/Magnesium (0.001) %/Oxygen (0.001) %/Silver (0.002) %/Cadmium (0.003) %/Chromium (0.003) %/Cobalt (0.003) %/Silicon (0.003) %/Antimony (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction