missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Kovar, Grade F15, K94610, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Stainless Steel, Kovar, Grade F15, K94610, 100g
Specifications
Specifications
| Chemical Name or Material | Carbon/Aluminium/Copper/Molybdenum/Silicon/Chromium/Manganese/Tin/Iron/Phosphorus/Tungsten/Cobalt/Nickel/Niobium/Titanium/Vanadium/Zirconium |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Carbon 0.019 %/Aluminium 0.04 %/Copper 0.05 %/Molybdenum 0.05 %/Silicon 0.1 %/Chromium 0.12 %/Manganese 0.24 %/Tin (<0.01) %/Iron (52.77) %/Phosphorus [0.002] %/Tungsten [0.03] %/Cobalt [17.50] %/Nickel [29.09] %/Niobium <0.01 %/Titanium <0.01 %/Vanadium <0.01 %/Zirconium <0.01 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction