missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI S7, T41907, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Tool Steel, AISI S7, T41907, 100g
Specifications
Specifications
| Chemical Name or Material | Lead/Zinc/Titanium/Antimony/Sulfur/Arsenic/Niobium/Tin/Cobalt/Aluminium/Tungsten/Phosphorus/Copper/Nickel/Vanadium/Manganese/Silicon/Carbon/Molybdenum/Chromium/Hydrogen/Cadmium/Tantalum/Magnesium/Beryllium/Zirconium/Bismuth/Boron/Oxygen/Nitrogen/Iron |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Lead 0.0003 %/Zinc 0.0011 %/Titanium 0.0014 %/Antimony 0.0016 %/Sulfur 0.0032 %/Arsenic 0.005 %/Niobium 0.005 %/Tin 0.006 %/Cobalt 0.0106 %/Aluminium 0.015 %/Tungsten 0.016 %/Phosphorus 0.021 %/Copper 0.128 %/Nickel 0.17 %/Vanadium 0.233 %/Manganese 0.271 %/Silicon 0.47 %/Carbon 0.51 %/Molybdenum 1.39 %/Chromium 3.28 %/Hydrogen (<0.0001) %/Cadmium (<0.0010) %/Tantalum (<0.0020) %/Magnesium (<0.0030) %/Beryllium (<0.0050) %/Zirconium (<0.0060) %/Bismuth (<0.0150) %/Boron (0.0003) %/Oxygen (0.0023) %/Nitrogen (0.0102) %/Iron (93.6) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction