missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Atorvastatin Calcium Salt, TRC

Product Code. 30005610
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005610 10 mg 10mg
30006831 5 mg 5mg
30009527 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005610 Supplier Toronto Research Chemicals Supplier No. TRCH82893010MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 265989-46-6
Molecular Formula C66H68CaF2N4O12
Molecular Weight (g/mol) 1187.35
Synonym (βR,δR)-2-(4-Fluorophenyl)-β,δ-dihydroxy-4-[[(2-hydroxyphenyl)amino]carbonyl]-5-(1-methylethyl)-3-phenyl-1H-pyrrole-1-heptanoic Acid Calcium Salt (2:1)
IUPAC Name calcium;(3R,5R)-7-[2-(4-fluorophenyl)-4-[(2-hydroxyphenyl)carbamoyl]-3-phenyl-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
SMILES FC(C=C1)=CC=C1C2=C(C3=CC=CC=C3)C(C(NC4=CC=CC=C4O)=O)=C(C(C)C)N2CC[C@@H](O)C[C@@H](O)CC([O-])=O.[Ca+2]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/2C33H35FN2O6.Ca/c2*1-20(2)31-30(33(42)35-26-10-6-7-11-27(26)39)29(21-8-4-3-5-9-21)32(22-12-14-23(34)15-13-22)36(31)17-16-24(37)18-25(38)19-28(40)41;/h2*3-15,20,24-25,37-39H,16-19H2,1-2H3,(H,35,42)(H,40,41);/q;;+2/p-2/t2*24-,25-;/m11./s1
Formula Weight 1186.4428
Percent Purity >95%
Chemical Name or Material 2-Hydroxy Atorvastatin Calcium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.