missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptidine Sulfate Salt, TRC
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
Specifications
Specifications
| Chemical Name or Material | Streptidine Sulfate Salt |
| Molecular Formula | C8H18N6O4 xH2SO4 |
| Quantity | 10 mg |
| Synonym | N1,N3-Bis(aminoiminomethyl)streptamine Sulfate Salt, N,N'-Diamidinostreptamine Sulfate Salt, N,N'''-(2,4,5,6-Tetrahydroxy-1,3-cyclohexanediyl)bis-guanidine Sulfate Salt |
| SMILES | O[C@H]1[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@@H](NC(N)=N)[C@@H]1O |
| Molecular Weight (g/mol) | 262.27 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction