Learn More
Fisherbrand™ 13mm Crimp Seal, Silver, Centre Tear-Off, assembled septum
Aluminum crimp closures provide a secure leak-resistant seal.
Brand: Fisherbrand™
Additional Details : Weight : 0.06000kg
Description
- Pre-assembled caps and septa are convenient and minimize contamination from handling
- Aluminum seals must be applied with a crimping tool
Specifications
PTFE/Butyl | |
13 mm | |
HPLC | |
2 mm |
Gray, Red | |
Aluminum | |
100 | |
Crimp Cap |