missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99491-100ul
This item is not returnable.
View return policy
Description
14-3-3 Polyclonal antibody specifically detects 14-3-3 in Human samples. It is validated for Western Blot
Specifications
| 14-3-3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human 14-3-3. | |
| 100 μg | |
| Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Protein Kinase, Signal Transduction | |
| 10971 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction