missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 227.00 - € 307.65

Specifications

Antigen 14-3-3
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18645155
View Documents
Novus Biologicals
NBP2-99491-100ul
100 μg
ONLINE EXCLUSIVE PRICE
€ 326.00
€ 307.65 / 100µL
Save € 18.35
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18642856
View Documents
Novus Biologicals
NBP2-99491-50ul
50 μg
€ 227.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

14-3-3 Polyclonal antibody specifically detects 14-3-3 in Human samples. It is validated for Western Blot
Specifications

Specifications

14-3-3
Western Blot
Unconjugated
Rabbit
Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Protein Kinase, Signal Transduction
0.2 um filtered solution in PBS
10971
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human
14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human 14-3-3.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.