missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron, Commercially Pure Iron, K00095, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Iron, Commercially Pure Iron, K00095, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Cobalt/Aluminium/Arsenic/Carbon/Manganese/Phosphorus/Oxygen/Copper/Chromium/Nickel/Silicon/Titanium/Tungsten/Zinc/Bismuth/Antimony/Selenium/Tantalum/Calcium/Magnesium/Nitrogen/Boron/Cadmium/Tin/Vanadium/Lead/Molybdenum/Niobium/Zirconium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cobalt 0.0009 %/Aluminium 0.0013 %/Arsenic 0.0016 %/Carbon 0.003 %/Manganese 0.003 %/Phosphorus 0.003 %/Oxygen 0.025 %/Copper 0.04 %/Chromium 0.043 %/Nickel 0.045 %/Silicon 0.07 %/Titanium (<0.005) %/Tungsten (<0.005) %/Zinc (<0.005) %/Bismuth (<0.007) %/Antimony (<0.01) %/Selenium (<0.01) %/Tantalum (<0.02) %/Calcium (0.0002) %/Magnesium (0.0002) %/Nitrogen (0.0003) %/Boron (0.0006) %/Cadmium (0.001) %/Tin (0.001) %/Vanadium (0.001) %/Lead (0.002) %/Molybdenum (0.002) %/Niobium (0.002) %/Zirconium (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction