missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Resulfurized, AISI 1118, G11180, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, Resulfurized, AISI 1118, G11180, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Lead/Vanadium/Oxygen/Arsenic/Tin/Cobalt/Nitrogen/Phosphorus/Aluminium/Molybdenum/Chromium/Carbon/Copper/Nickel/Silicon/Manganese/Calcium/Cadmium/Zinc/Antimony/Boron/Tantalum/Magnesium/Selenium/Bismuth/Niobium/Zirconium/Tungsten |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium 0.0015 %/Lead 0.002 %/Vanadium 0.0028 %/Oxygen 0.003 %/Arsenic 0.008 %/Tin 0.009 %/Cobalt 0.01 %/Nitrogen 0.0108 %/Phosphorus 0.0113 %/Aluminium 0.032 %/Molybdenum 0.045 %/Chromium 0.104 %/Carbon 0.163 %/Copper 0.19 %/Nickel 0.197 %/Silicon 0.281 %/Manganese 1.44 %/Calcium (<0.0005) %/Cadmium (<0.001) %/Zinc (<0.002) %/Antimony (<0.004) %/Boron (<0.005) %/Tantalum (<0.005) %/Magnesium (<0.006) %/Selenium (<0.006) %/Bismuth (<0.02) %/Niobium (0.002) %/Zirconium (0.002) %/Tungsten (0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction