missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Electrolytic Tough Pitch, CDA 110, C11000, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Electrolytic Tough Pitch, CDA 110, C11000, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Tin/Silicon/Arsenic/Silver/Lead/Aluminium/Cobalt/Phosphorus/Iron/Carbon/Copper/Indium/Germanium/Beryllium/Nitrogen/Chromium/Manganese/Magnesium/Boron/Nickel/Cadmium/Selenium/Tellurium/Zirconium/Bismuth/Oxygen/Zinc/Antimony/Niobium |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tin 0.0005 %/Silicon 0.0006 %/Arsenic 0.0009 %/Silver 0.0011 %/Lead 0.0013 %/Aluminium 0.0014 %/Cobalt 0.0014 %/Phosphorus 0.0014 %/Iron 0.0016 %/Carbon 0.002 %/Copper 99.94 %/Indium (<0.0001) %/Germanium (<0.0002) %/Beryllium (<0.002) %/Nitrogen (<0.008) %/Chromium (0.0002) %/Manganese (0.0002) %/Magnesium (0.0003) %/Boron (0.0004) %/Nickel (0.0004) %/Cadmium (0.0008) %/Selenium (0.001) %/Tellurium (0.001) %/Zirconium (0.001) %/Bismuth (0.002) %/Oxygen (0.002) %/Zinc (0.002) %/Antimony (0.003) %/Niobium (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction