missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Silver Zirconium, NARLOY-Z, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Silver Zirconium, NARLOY-Z, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Carbon/Zirconium/Silver/Copper/Nitrogen/Oxygen/Phosphorus/Aluminium/Chromium/Cobalt/Iron/Lead/Manganese/Nickel/Silicon/Tin/Zinc |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Carbon 0.003 %/Zirconium 0.04 %/Silver 3.03 %/Copper 96.6 %/Nitrogen [0.0002] %/Oxygen [0.0030] %/Phosphorus [0.004] %/Aluminium <0.01 %/Chromium <0.01 %/Cobalt <0.01 %/Iron <0.01 %/Lead <0.01 %/Manganese <0.01 %/Nickel <0.01 %/Silicon <0.01 %/Tin <0.01 %/Zinc <0.01 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction