missing translation for 'onlineSavingsMsg'
Learn More

Copper, Phosphor Bronze 5% A, CDA 510, C51000, 100g, ARMI∣MBH

Product Code. 17977984
Change view
Click to view available options
Quantity:
100 g
Unit Size:
Each
Product Code. Quantity unitSize
17977984 100 g Each
1 options
This item is not returnable. View return policy

Product Code. 17977984

Brand: ARMI/MBH IARMCU51018C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Copper, Phosphor Bronze 5% A, CDA 510, C51000, 100g
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxygen/Carbon/Silver/Lead/Zinc/Nickel/Phosphorus/Tin/Copper/Aluminium/Antimony/Arsenic/Bismuth/Boron/Cadmium/Calcium/Chromium/Cobalt/Hydrogen/Iron/Magnesium/Manganese/Molybdenum/Nitrogen/Palladium/Selenium/Silicon/Sulfur/Tellurium/Titanium/Tungsten/Vanadium
CAS 7440-50-8
Quantity 100 g
Form Pieces
Concentration or Composition (by Analyte or Components) Oxygen 0.0011 %/Carbon 0.0018 %/Silver 0.0019 %/Lead 0.0027 %/Zinc 0.0034 %/Nickel 0.017 %/Phosphorus 0.16 %/Tin 4.54 %/Copper 95.2 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.