missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Zn/Al, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Zinc, Zn/Al, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Manganese/Lead/Lanthanum/Cerium/Cadmium/Copper/Zirconium/Titanium/Iron/Magnesium/Aluminium/Nickel/Tin/Silicon |
| CAS | 7440-66-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Manganese 0.0006 %/Lead 0.0021 %/Lanthanum 0.0044 %/Cerium 0.0047 %/Cadmium 0.0054 %/Copper 0.007 %/Zirconium 0.011 %/Titanium 0.02 %/Iron 0.032 %/Magnesium 0.0464 %/Aluminium 5.58 %/Nickel (0.0003) %/Tin (0.00035) %/Silicon (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction