missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Galvanizing, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Zinc, Galvanizing, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Arsenic/Cobalt/Magnesium/Nickel/Bismuth/Copper/Cadmium/Aluminium/Lead/Tin/Iron/Antimony |
| CAS | 7440-66-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Arsenic 0.00044 %/Cobalt 0.0011 %/Magnesium 0.0014 %/Nickel 0.0025 %/Bismuth 0.0098 %/Copper 0.0103 %/Cadmium 0.0136 %/Aluminium 0.0139 %/Lead 0.0166 %/Tin 0.0172 %/Iron 0.0443 %/Antimony 0.148 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction