missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASRegistryNumber1t Iron, Pig Iron Medium, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
CASRegistryNumber1t Iron, Pig Iron Medium, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Lead/Arsenic/Zinc/Calcium/Boron/Zirconium/Tungsten/Tin/Copper/Niobium/Cobalt/Molybdenum/Nickel/Phosphorus/Sulfur/Chromium/Aluminium/Vanadium/Manganese/Titanium/Silicon/Carbon/Iron |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead -40 ppm/Arsenic -18 ppm/Zinc -17 ppm/Calcium -11 ppm/Boron -4 ppm/Zirconium 0.0022 %/Tungsten 0.003 %/Tin 0.0061 %/Copper 0.0117 %/Niobium 0.014 %/Cobalt 0.015 %/Molybdenum 0.0157 %/Nickel 0.028 %/Phosphorus 0.051 %/Sulfur 0.053 %/Chromium 0.057 %/Aluminium 0.06 %/Vanadium 0.071 %/Manganese 0.077 %/Titanium 0.258 %/Silicon 0.71 %/Carbon 3.22 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction