missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Residuals, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Aluminum, Residuals, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Zirconium/Lead/Chromium/Manganese/Vanadium/Copper/Titanium/Bismuth/Magnesium/Tin/Silicon/Nickel/Zinc/Iron/Cadmium/Antimony |
| CAS | 7429-90-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Zirconium 0.0008 %/Lead 0.0021 %/Chromium 0.0062 %/Manganese 0.0074 %/Vanadium 0.008 %/Copper 0.0132 %/Titanium 0.0135 %/Bismuth 0.024 %/Magnesium 0.054 %/Tin 0.097 %/Silicon 0.143 %/Nickel 0.23 %/Zinc 0.249 %/Iron 0.835 %/Cadmium (0.0001) %/Antimony (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction