missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Residuals, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Residuals, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Gold/Indium/Cadmium/Germanium/Cobalt/Arsenic/Magnesium/Nickel/Aluminium/Chromium/Selenium/Phosphorus/Zinc/Manganese/Iron/Tellurium/Antimony/Bismuth/Silver/Tin/Lead |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Gold (0.0022) %/Indium (0.0025) %/Cadmium (0.0026) %/Germanium (0.004) %/Cobalt (0.008) %/Arsenic (0.01) %/Magnesium (0.01) %/Nickel (0.011) %/Aluminium (0.018) %/Chromium (0.018) %/Selenium (0.021) %/Phosphorus (0.025) %/Zinc (0.025) %/Manganese (0.032) %/Iron (0.0365) %/Tellurium (0.038) %/Antimony (0.039) %/Bismuth (0.04) %/Silver (0.04) %/Tin (0.069) %/Lead (0.07) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction