missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Residuals, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Residuals, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Aluminium/Magnesium/Germanium/Selenium/Tellurium/Iron/Silver/Antimony/Indium/Bismuth/Silicon/Phosphorus/Manganese/Cobalt/Chromium/Cadmium/Arsenic/Lead/Nickel/Tin/Zinc/Gold |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.0028 %/Magnesium 0.0041 %/Germanium 0.0052 %/Selenium 0.0053 %/Tellurium 0.0065 %/Iron 0.0075 %/Silver 0.0086 %/Antimony 0.0095 %/Indium 0.0097 %/Bismuth 0.0109 %/Silicon 0.0111 %/Phosphorus 0.0125 %/Manganese 0.0147 %/Cobalt 0.0273 %/Chromium 0.0316 %/Cadmium 0.041 %/Arsenic 0.0435 %/Lead 0.0501 %/Nickel 0.0511 %/Tin 0.0614 %/Zinc 0.084 %/Gold (0.0004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction