missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI S1, T41901, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Tool Steel, AISI S1, T41901, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Boron/Oxygen/Niobium/Nitrogen/Titanium/Aluminium/Cobalt/Tin/Phosphorus/Nickel/Copper/Vanadium/Molybdenum/Manganese/Carbon/Silicon/Chromium/Tungsten/Arsenic/Lead |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0003 %/Oxygen 0.002 %/Niobium 0.003 %/Nitrogen 0.0069 %/Titanium 0.007 %/Aluminium 0.011 %/Cobalt 0.013 %/Tin 0.016 %/Phosphorus 0.019 %/Nickel 0.108 %/Copper 0.147 %/Vanadium 0.17 %/Molybdenum 0.222 %/Manganese 0.27 %/Carbon 0.45 %/Silicon 0.89 %/Chromium 1.09 %/Tungsten 1.96 %/Arsenic (0.01) %/Lead <0.002 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction