Learn More
2-(1-Pyrrolidinyl)nicotinaldehyde, 97%, Thermo Scientific™
Product Code. 10269052
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269052 250 mg Amber Glass Bottle 250mg
10300883 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10269052 Supplier Thermo Scientific Maybridge Supplier No. MO07059CB
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Chemical Identifiers
CAS 690632-39-4
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.22
MDL Number MFCD05865136
InChI Key FTACFSVJFQMXQE-UHFFFAOYSA-N
Synonym 2-1-pyrrolidinyl nicotinaldehyde, 2-pyrrolidin-1-yl nicotinaldehyde, 3-pyridinecarboxaldehyde,2-1-pyrrolidinyl, 2-pyrrolidin-1-yl pyridine-3-carbaldehyde, 2-pyrrolidinylpyridine-3-carbaldehyde, 2-pyrrolidin-1-yl-pyridine-3-carbaldehyde, 2-1-pyrrolidinyl-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 2-1-pyrrolidinyl
PubChem CID 2794763
IUPAC Name 2-pyrrolidin-1-ylpyridine-3-carbaldehyde
SMILES O=CC1=C(N=CC=C1)N1CCCC1
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 176.22
Percent Purity 97%
Chemical Name or Material 2-(1-Pyrrolidinyl)nicotinaldehyde