Learn More
2-(1H-Pyrazol-1-yl)pyridine, Thermo Scientific™
Product Code. 17505595 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17505595 1 g 1g
17515595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17505595 Supplier Thermo Scientific Acros Supplier No. 469560010
Request Bulk or Custom Format
Chemical Identifiers
CAS 25700-11-2
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number 09033534
InChI Key XXTPHXNBKRVYJI-UHFFFAOYSA-N
IUPAC Name 2-(1H-pyrazol-1-yl)pyridine
SMILES C1=CN(N=C1)C1=CC=CC=N1
Specifications
Alpha Vector PYRAZOLYLPYRIDINE
Melting Point 35°C to 45°C
Color White to almost white
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 1 g
Solubility Information Soluble in methanol
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-(1H-Pyrazol-1-yl)pyridine