Learn More
2-(2-Cyclohexyl-2-hydroxy-2-phenylacetyloxy)acetic Acid, TRC
Product Code. 30350108
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350108 1 g -
30352157 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30350108 Supplier Toronto Research Chemicals Supplier No. TRCC9880801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1797008-83-3
Molecular Formula C16H20O5
Molecular Weight (g/mol) 292.33
IUPAC Name 2-(2-cyclohexyl-2-hydroxy-2-phenylacetoxy)acetic acid
SMILES OC(COC(C(C1CCCCC1)(O)C2=CC=CC=C2)=O)=O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C16H20O5/c17-14(18)11-21-15(19)16(20,12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1,3-4,7-8,13,20H,2,5-6,9-11H2,(H,17,18)
Formula Weight 292.13
Chemical Name or Material 2-(2-Cyclohexyl-2-hydroxy-2-phenylacetyloxy)acetic Acid