Learn More
2,3,4,5,6-Pentafluorotoluene, 99%
Product Code. p-7025759
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11489387
Brand: Thermo Scientific Alfa Aesar B21419.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 771-56-2 | C7H3F5 | 182.093 g/mol
2,3,4,5,6-Pentafluorotoluene, can be used as an organic fluorinated building blocks for chemical synthesis. It is also used as an pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,4,5,6-Pentafluorotoluene, can be used as an organic fluorinated building blocks for chemical synthesis. It is also used as an pharmaceutical intermediate.
Solubility
It is soluble in most common organic solvents and insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 771-56-2
Molecular Formula C7H3F5
Molecular Weight (g/mol) 182.093
MDL Number MFCD00000298
InChI Key SXPRVMIZFRCAGC-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorotoluene, pentafluorotoluene, methyl pentafluorobenzene, pentafluoromethylbenzene, benzene, pentafluoromethyl, methylpentafluorobenzene, benzene, 1,2,3,4,5-pentafluoro-6-methyl, toluene, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro-toluene, n-methylpentafluorobenzene
PubChem CID 69869
IUPAC Name 1,2,3,4,5-pentafluoro-6-methylbenzene
SMILES CC1=C(C(=C(C(=C1F)F)F)F)F
Specifications
Melting Point -30°C
Density 1.439
Boiling Point 116°C to 117°C
Flash Point 34°C (93°F)
Refractive Index 1.403
Quantity 10 g
UN Number UN1993
Beilstein 1911885
Solubility Information It is soluble in most common organic solvents and insoluble in water.
Formula Weight 182.09
Percent Purity 99%
Chemical Name or Material 2,3,4,5,6-Pentafluorotoluene