Learn More
2,3-Butanediol, TRC
Specifications
Specifications
| CAS | 513-85-9 |
| Molecular Formula | C4 H10 O2 |
| Quantity | 10 g |
| Synonym | 1,2-Dimethyl-1,2-ethanediol, 2,3-Butylene glycol, 2,3-Dihydroxybutane, Dimethylethylene glycol |
| InChI Formula | InChI=1S/C4H10O2/c1-3(5)4(2)6/h3-6H,1-2H3 |
| SMILES | CC(O)C(C)O |
| IUPAC Name | butane-2,3-diol |
| Molecular Weight (g/mol) | 90.12 |
| Formula Weight | 90.0681 |
| Chemical Name or Material | 2,3-Butanediol |