missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Methyl-2-furyl)ethanamine, TRC

Product Code. 30470543
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470543 1 g 1g
30470661 100 mg 100mg
30481366 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470543 Supplier Toronto Research Chemicals Supplier No. TRCM3280581G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30796-85-1
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.16
Synonym 5-Methyl-2-furanethanamine, 2-(5-Methylfuran-2-yl)ethan-1-amine, 2-(5-Methylfuran-2-yl)ethanamine, 2-(5-Methylfuran-2-yl)ethylamine, beta-(5-Methyl-2-furanyl)ethanamine
IUPAC Name 2-(5-methylfuran-2-yl)ethanamine
SMILES Cc1oc(CCN)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H11NO/c1-6-2-3-7(9-6)4-5-8/h2-3H,4-5,8H2,1H3
Formula Weight 125.0841
Chemical Name or Material 2-(5-Methyl-2-furyl)ethanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.