Learn More
2-Acetyl-5-chlorothiophene, 99%
Product Code. p-7081744
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11453814
Brand: Thermo Scientific Alfa Aesar A11850.09
Request Bulk or Custom Format
CAS: 6310-09-4 | C6H5ClOS | 160.62 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6310-09-4
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00005444
InChI Key HTZGPEHWQCRXGZ-UHFFFAOYSA-N
Synonym 2-acetyl-5-chlorothiophene, 1-5-chlorothiophen-2-yl ethanone, 1-5-chloro-2-thienyl ethanone, 5-chloro-2-acetylthiophene, ethanone, 1-5-chloro-2-thienyl, 5-chlor-2-acetyl thiophen, ketone, 5-chloro-2-thienyl methyl, 1-5-chlorothiophen-2-yl ethan-1-one, 5-chloro-2-thienyl methyl ketone, 2-chloro-5-acetylthiophene
PubChem CID 80572
IUPAC Name 1-(5-chlorothiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(S1)Cl
Specifications
Melting Point 44°C to 48°C
Boiling Point 117°C to 118°C (17 mmHg)
Flash Point 108°C (226°F)
Quantity 10 g
UN Number UN2811
Beilstein 113936
Sensitivity Light sensitive
Formula Weight 160.62
Percent Purity 99%
Chemical Name or Material 2-Acetyl-5-chlorothiophene