missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-6-isopropylpyrimidine, 97%, Thermo Scientific™

Product Code. 11361899
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361899 1 g 1g
15493327 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361899 Supplier Thermo Scientific Alfa Aesar Supplier No. H25997.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Amino-4-chloro-6-isopropylpyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73576-33-7
Molecular Formula C7H10ClN3
Molecular Weight (g/mol) 171.63
MDL Number MFCD03840198
InChI Key SPMMDFHRMVXLLO-UHFFFAOYSA-N
Synonym 4-chloro-6-isopropylpyrimidin-2-amine, 2-amino-4-chloro-6-isopropylpyrimidine, 6-chloro-4-methylethyl pyrimidine-2-ylamine, 4-chloro-6-propan-2-yl pyrimidin-2-amine, 4-chloro-6-isopropyl-pyrimidin-2-amine, 4-chloro-6-isopropyl-2-pyrimidinamine, 2-pyrimidinamine,4-chloro-6-1-methylethyl, 4-chloro-6-prop-2-yl pyrimidin-2-amine, 4-chloro-6-isopropyl-pyrimidin-2-ylamine, 4-chloro-6-1-methylethyl pyrimidin-2-amine
PubChem CID 2773619
IUPAC Name 4-chloro-6-propan-2-ylpyrimidin-2-amine
SMILES CC(C)C1=CC(Cl)=NC(N)=N1

Specifications

Melting Point 98°C to 101°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 171.63
Percent Purity 97%
Chemical Name or Material 2-Amino-4-chloro-6-isopropylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.