missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzylmethylamino)-3',4'-dihydroxyacetophenone, TRC

Product Code. 30003699
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003699 10 g 10g
30007101 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003699 Supplier Toronto Research Chemicals Supplier No. TRCB28562010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36467-25-1
Molecular Formula C16 H17 N O3
Molecular Weight (g/mol) 271.31
Synonym Acetophenone, 2-(benzylmethylamino)-3',4'-dihydroxy- (6CI), 1-(3,4-Dihydroxyphenyl)-2-[methyl(phenylmethyl)amino]ethanone, α-Benzylmethylamino-3,4-dihydroxyacetophenone, 2-(Benzylmethylamino)-1-(3,4-dihydroxyphenyl)ethanone, Adrenaline Tartrate Imp. E (EP)
IUPAC Name 2-[benzyl(methyl)amino]-1-(3,4-dihydroxyphenyl)ethanone
SMILES CN(CC(=O)c1ccc(O)c(O)c1)Cc2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H17NO3/c1-17(10-12-5-3-2-4-6-12)11-16(20)13-7-8-14(18)15(19)9-13/h2-9,18-19H,10-11H2,1H3
Formula Weight 271.1208
Percent Purity >95%
Chemical Name or Material 2-(Benzylmethylamino)-1-(3,4-dihydroxyphenyl)ethanone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.