Learn More
2-Bromo-1-[4-(3-bromothien-2-yl)phenyl]ethanone, 97%, Thermo Scientific™
Product Code. 10662505
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10662505 250 mg Amber Glass Bottle 250mg
10664015 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10662505 Supplier Thermo Scientific Maybridge Supplier No. CC67805CB
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 944450-89-9
Molecular Formula C12H8Br2OS
Molecular Weight (g/mol) 360.06
MDL Number MFCD09879969
InChI Key YTVAARIMMJZZIE-UHFFFAOYSA-N
Synonym 2-bromo-1-4-3-bromothien-2-yl phenyl ethanone, 2-bromo-1-4-3-bromothiophen-2-yl phenyl ethan-1-one, 2-bromo-1-4-3-bromothiophen-2-yl phenyl ethanone, 4-3-bromothien-2-yl phenacyl bromide, 4-4-bromothien-2-yl phenacyl bromide
PubChem CID 24229752
IUPAC Name 2-bromo-1-[4-(3-bromothiophen-2-yl)phenyl]ethanone
SMILES BrCC(=O)C1=CC=C(C=C1)C1=C(Br)C=CS1
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 360.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-[4-(3-bromothien-2-yl)phenyl]ethanone