missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-chlorophenyl)propan-1-one, TRC

Product Code. 30133307
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133307 250 mg 250mg
30134388 500 mg 500mg
30009403 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133307 Supplier Toronto Research Chemicals Supplier No. TRCB684293250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-37-2
Molecular Formula C9H8BrClO
Molecular Weight (g/mol) 247.52
IUPAC Name 2-bromo-1-(4-chlorophenyl)propan-1-one
SMILES CC(Br)C(=O)c1ccc(Cl)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H8BrClO/c1-6(10)9(12)7-2-4-8(11)5-3-7/h2-6H,1H3
Formula Weight 245.9447
Chemical Name or Material 2-Bromo-1-(4-chlorophenyl)propan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.