Learn More
2-Bromo-1-(4-isobutylphenyl)propan-1-one, 97%, Thermo Scientific™
Product Code. 10446672
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10446672 1 g Amber Glass Bottle 1g
10682263 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10446672 Supplier Thermo Scientific Maybridge Supplier No. RH01005DA
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 80336-64-7
Molecular Formula C13H17BrO
Molecular Weight (g/mol) 269.18
MDL Number MFCD00831700
InChI Key SVGMFVCPJFHTRE-UHFFFAOYNA-N
Synonym 2-bromo-1-4-isobutylphenyl propan-1-one, 2-bromo-1-4-2-methylpropyl phenyl propan-1-one, 1-4-isobutylphenyl-2-bromo-1-propanone, 2-bromo-4'-2-methylprop-1-yl propiophenone, 2-bromo-1-4-2-methylpropyl phenyl-1-propanone, 2-bromanyl-1-4-2-methylpropyl phenyl propan-1-one, 1-propanone,2-bromo-1-4-1,1-dimethylethyl phenyl
PubChem CID 2735576
IUPAC Name 2-bromo-1-[4-(2-methylpropyl)phenyl]propan-1-one
SMILES CC(C)CC1=CC=C(C=C1)C(=O)C(C)Br
Specifikationer
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3CH(Br)C(O)C6H4CH2CH(CH3)2
Quantity 1 g
Formula Weight 269.18
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(4-isobutylphenyl)propan-1-one
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.