missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-dichloroacetophenone, 95%

Product Code. p-7040029
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425998 1 g 1g
15435998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425998 Supplier Thermo Scientific Alfa Aesar Supplier No. H59615.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2631-72-3 | C8H5BrCl2O | 267.931 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2631-72-3
Molecular Formula C8H5BrCl2O
Molecular Weight (g/mol) 267.931
MDL Number MFCD00053005
InChI Key DASJDMQCPIDJIF-UHFFFAOYSA-N
Synonym 2-bromo-2',4'-dichloroacetophenone, 2-bromo-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl bromide, 2-bromo-1-2,4-dichlorophenyl ethan-1-one, 2-bromo-1-2,4-dichloro-phenyl-ethanone, ethanone, 2-bromo-1-2,4-dichlorophenyl, 2'-bromo-2,4-dichloro acetophenone, omega-bromo-2,4-dichloro acetophenone, 2',4'-dichlorophenacyl bromide, 2-bromo-2,4-dichloroacetophenone
PubChem CID 75828
IUPAC Name 2-bromo-1-(2,4-dichlorophenyl)ethanone
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)CBr

Specifications

Melting Point 25°C to 29°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN1759
Formula Weight 267.93
Percent Purity 95%
Chemical Name or Material 2-Bromo-2',4'-dichloroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.