Learn More
2-Chloro-4-iodo-3-methylpyridine, 95%
Product Code. 11494540
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494540 1 g 1g
11404550 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11494540 Supplier Thermo Scientific Alfa Aesar Supplier No. H50107.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 153034-88-9 | C6H5ClIN | 253.467 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 153034-88-9
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.467
MDL Number MFCD03095293
InChI Key WHCRABJNJRAJIP-UHFFFAOYSA-N
Synonym 2-chloro-4-iodo-3-picoline, 2-chloro-3-methyl-4-iodopyridine, 2-chloro-4-iodo-3-methyl-pyridine, pyridine, 2-chloro-4-iodo-3-methyl, 2-chloro-4-iodo-3-methylpyridne, pubchem17059, 2-chloranyl-4-iodanyl-3-methyl-pyridine, 2-chloro-4-iodo-3-methylpyridine, 2-chloro-4-iodo-3-methyl-pyridine; 2-chloro-4-iodo-3-picoline
PubChem CID 10083628
IUPAC Name 2-chloro-4-iodo-3-methylpyridine
SMILES CC1=C(C=CN=C1Cl)I
Specifications
Melting Point 106°C to 107°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 253.47
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-iodo-3-methylpyridine