missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™

Product Code. 11336919
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
This item is not returnable. View return policy

Product Code. 11336919

Brand: Thermo Scientific Alfa Aesar H26805.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 280566-45-2
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.551
MDL Number MFCD01862658
InChI Key DXRBTBMFFGEVCX-UHFFFAOYSA-N
Synonym 2-chloro-6-trifluoromethyl nicotinic acid, 2-chloro-6-trifluoromethylnicotinic acid, 2-chloro-6-trifluoromethyl pyridine-3-carboxylic acid, 2-chloro-6-trifluoromethyl-nicotinic acid, 2-chloro-6-trifluoromethyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro-6-trifluoromethyl, pubchem13677, acmc-1c67o, 2-chloro-6-trifluoromethyl nicotinicacid
PubChem CID 2778210
IUPAC Name 2-chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES C1=CC(=NC(=C1C(=O)O)Cl)C(F)(F)F

Specifications

Melting Point 118°C to 120°C
Quantity 500 mg
Formula Weight 225.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)nicotinic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.