missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-6-fluoropyridine, 95%

Product Code. 11446520
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11446520

Brand: Thermo Scientific Alfa Aesar H27600.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3939-15-9 | C6H3FN2 | 122.10 g/mol

It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3939-15-9
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.10
MDL Number MFCD09263323
InChI Key NVOLTPVZQXTZCW-UHFFFAOYSA-N
Synonym 2-cyano-6-fluoropyridine, 6-fluoropicolinonitrile, 2-pyridinecarbonitrile, 6-fluoro, 6-fluoro-2-cyanopyridine;2-fluoro-6-cyanopyridine;6-fluoropicolinonitrile, pubchem4017, acmc-1cj0q, 2-pyridinecarbonitrile,6-fluoro, 6-fluoro-pyridine-2-carbonitrile, 2-fluoropyridine-6-carbonitrile, 6-fluoro-2-pyridinecarbonitrile
PubChem CID 14296369
IUPAC Name 6-fluoropyridine-2-carbonitrile
SMILES FC1=CC=CC(=N1)C#N

Specifications

Melting Point 32°C to 35°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 122.1
Percent Purity 95%
Chemical Name or Material 2-Cyano-6-fluoropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.