Learn More
2-Fluoro-4-methylpyridine, 98%
Product Code. 11311347
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311347 5 g 5g
11301347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11311347 Supplier Thermo Scientific Alfa Aesar Supplier No. L01602.06
Request Bulk or Custom Format
CAS: 461-87-0 | C6H6FN | 111.119 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 461-87-0
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD00041224
InChI Key ZBFAXMKJADVOGH-UHFFFAOYSA-N
Synonym 2-fluoro-4-picoline, 2-fluoro-4-methyl-pyridine, pyridine, 2-fluoro-4-methyl, 2-fluoro-4-methyl pyridine, pubchem1203, acmc-1aevi, 4-methyl-2-fluoropyridine, 2-fluoro-4-methylpyridine,, ksc494a2j
PubChem CID 2737403
IUPAC Name 2-fluoro-4-methylpyridine
SMILES CC1=CC(=NC=C1)F
Specifications
Density 1.078
Boiling Point 155°C to 156°C
Flash Point 51°C (123°F)
Refractive Index 1.4725
Quantity 5 g
UN Number UN1993
Beilstein 107086
Formula Weight 111.12
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-methylpyridine