Learn More
2-Hydroxy Hippuric Acid-13C2,15N, TRC
Product Code. 30179874
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179874 1 mg 1mg
30180084 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30179874 Supplier Toronto Research Chemicals Supplier No. TRCH9428871MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1286521-95-6
Molecular Formula C713C2H915NO4
Molecular Weight (g/mol) 198.15
Synonym N-(2-Hydroxybenzoyl)glycine-13C2,15N, o-Hydroxy-hippuric Acid, Salicyluric Acid-13C2,15N, (2-Hydroxybenzoyl)glycine-13C2,15N, 2-Hydroxyhippuric Acid-13C2,15N, NSC 524135-13C2,15N, Salicyloylglycine-13C2,15N, o-Hydroxyhippuric Acid-13C2,15N;
IUPAC Name 2-[(2-hydroxybenzoyl)amino]acetic acid
SMILES OC1=CC=CC=C1C([15NH][13CH2][13C](O)=O)=O
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C9H9NO4/c11-7-4-2-1-3-6(7)9(14)10-5-8(12)13/h1-4,11H,5H2,(H,10,14)(H,12,13)/i5+1,8+1,10+1
Formula Weight 198.0569
Chemical Name or Material 2-Hydroxy Hippuric Acid-13C2,15N