missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-(trifluoromethyl)aniline, 99%

Product Code. 11415337
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11415337

Brand: Thermo Scientific Alfa Aesar B22691.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 163444-17-5 | C7H5F3IN | 287.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163444-17-5
Molecular Formula C7H5F3IN
Molecular Weight (g/mol) 287.02
MDL Number MFCD02683097
InChI Key UKKWTZPXYIYONW-UHFFFAOYSA-N
Synonym 4-amino-3-iodobenzotrifluoride, 2-iodo-4-trifluoromethyl aniline, 4-trifluoromethyl-2-iodobenzenamine, benzenamine, 2-iodo-4-trifluoromethyl, 2-iodo-4-trifluoromethyl benzenamine, pubchem1632, acmc-209dpp, intermediates-zcf02228, ksc910s3b, 3-iodo-4-aminobenzotrifluoride
PubChem CID 2783322
IUPAC Name 2-iodo-4-(trifluoromethyl)aniline
SMILES NC1=C(I)C=C(C=C1)C(F)(F)F

Specifications

Melting Point 49°C to 52°C
Quantity 50 g
Sensitivity Light sensitive
Formula Weight 287.02
Percent Purity 99%
Chemical Name or Material 2-Iodo-4-(trifluoromethyl)aniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.