missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAMHD1 Recombinant Rabbit Monoclonal Antibody (24GB1010)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557701
This item is not returnable.
View return policy
Description
SAMHD1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene may play a role in regulation of the innate immune response. The encoded protein is upregulated in response to viral infection and may be involved in mediation of tumor necrosis factor-alpha proinflammatory responses. Mutations in this gene have been associated with Aicardi-Goutieres syndrome.
Specifications
| SAMHD1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SAMHD1 | |
| CHBL2; DCIP; Dendritic cell-derived IFNG-induced protein; deoxynucleoside triphosphate triphosphohydrolase SAMHD1; dNTPase; E330031J07Rik; EGK_02438; HDDC1; hSAMHD1; IFN-gamma induced; interferon-gamma-inducible protein Mg11; Mg11; Mg21; Monocyte protein 5; MOP5; MOP-5; mSAMHD1; RP1-132F21.1; SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1; SAM domain and HD domain 1; SAM domain and HD domain, 1; SAM domain and HD domain-containing protein 1; Samhd1; SBBI88 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25939 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1010 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y3Z3 | |
| SAMHD1 | |
| A synthesized peptide derived from human SAMHD1 (aa 580-626). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction