Description
This package helps achieve full compliance with Subpart B - Electronic Records and Subpart C - Electronic Signatures of the regulation:
- Track and record settings and any changes to settings with the system log function running permanently, serving as an audit trail
- Track who performs any operation as analysis results are automatically signed with the user information (each user has a unique identification and electronic signature)
- Ensure the quality and incorruptibility of recorded data with backup procedures (exported files are protected from unwanted or improper alteration)
- Ensure the right delegation of responsibilities at the right level with the user management system with three access levels
Specifications
Specifications
| Type | Software Package |
| For Use With (Equipment) | UDK 169 or 159 Distillation Units |