missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2A Peptide Antibody (3H4R) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-09171SS
This item is not returnable.
View return policy
Description
2A Peptide Monoclonal antibody specifically detects 2A Peptide in Human, Nonspecies specific samples. It is validated for Western Blot
Specifications
| 2A Peptide | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 μg/ml | |
| 2A cleavage peptide, 2A Peptide, 2A peptide tag, 2A sequence, 2A-like peptide, antibody for 2A-containing recombinant proteins, FMDV 2A-peptide, MDV 2A-peptide, P2A peptide, P2A tagged proteins, peptide 2A sequences, self-cleaving 2A peptide, T2A peptide, T2A tagged proteins, Thosea asigna virus 2A | |
| This Recombinant 2A Peptide Antibody (3H4) was developed against KLH-coupled synthetic peptide corresponding to 2A peptide of Thosea asigna virus (CGDVEENPG) | |
| 0.025 mg | |
| Epitope Tags, Virology Bacteria and Parasites | |
| Human, Non-species specific | |
| Purified |
| Western Blot | |
| 3H4R | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction