missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2A Peptide Antibody (3H4R) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 244.00 - € 506.00
Specifications
| Antigen | 2A Peptide |
|---|---|
| Clone | 3H4R |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 - 2 μg/ml |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616978
|
Novus Biologicals
NBP3-09171 |
0.1 mg |
€ 506.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676668
|
Novus Biologicals
NBP3-09171SS |
0.025 mg |
€ 244.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2A Peptide Monoclonal antibody specifically detects 2A Peptide in Human, Nonspecies specific samples. It is validated for Western BlotSpecifications
| 2A Peptide | |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Epitope Tags, Virology Bacteria and Parasites | |
| PBS | |
| This Recombinant 2A Peptide Antibody (3H4) was developed against KLH-coupled synthetic peptide corresponding to 2A peptide of Thosea asigna virus (CGDVEENPG) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 3H4R | |
| Western Blot 1 - 2 μg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Non-species specific | |
| 2A cleavage peptide, 2A Peptide, 2A peptide tag, 2A sequence, 2A-like peptide, antibody for 2A-containing recombinant proteins, FMDV 2A-peptide, MDV 2A-peptide, P2A peptide, P2A tagged proteins, peptide 2A sequences, self-cleaving 2A peptide, T2A peptide, T2A tagged proteins, Thosea asigna virus 2A | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title